siRNA Details
| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767175.1 | XP_008765397.1 | 21 | 3318 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nbeal1| Gene Symbol: Nbeal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767175.1 | XP_008765397.1 | 21 | 3318 | ||