siRNA Details
| Gene Symbol: Adam23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029899.1 | NP_001025070.1 | 11 | 1197 | ||
| XM_008767089.1 | XP_008765311.1 | 11 | 1458 | |||
| XM_008767090.1 | XP_008765312.1 | 11 | 1458 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam23| Gene Symbol: Adam23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029899.1 | NP_001025070.1 | 11 | 1197 | ||
| XM_008767089.1 | XP_008765311.1 | 11 | 1458 | |||
| XM_008767090.1 | XP_008765312.1 | 11 | 1458 | |||