siRNA Details
| Gene Symbol: Fastkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009673.1 | NP_001009673.1 | 10, 11 | 2024 | ||
| XM_006245050.1 | XP_006245112.1 | 11, 12 | 2181 | |||
| GenBank mRNA | BC088416.1 | - | 10, 11 | 2024 | 7112913 | |
| FQ218822.1 | - | 6, 7 | 829 | |||

Fastkd2
Genomic Map