siRNA Details
| Gene Symbol: Fastkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009673.1 | NP_001009673.1 | 1 | 396 | ||
| XM_006245050.1 | XP_006245112.1 | 2 | 523 | |||
| GenBank mRNA | BC088416.1 | - | 1 | 396 | 7112913 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fastkd2| Gene Symbol: Fastkd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009673.1 | NP_001009673.1 | 1 | 396 | ||
| XM_006245050.1 | XP_006245112.1 | 2 | 523 | |||
| GenBank mRNA | BC088416.1 | - | 1 | 396 | 7112913 | |