siRNA Details
| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009357.1 | NP_001009357.1 | 5 | 863 | ||
| GenBank mRNA | BC087134.1 | - | 5 | 863 | 7122272 | |
| FQ225078.1 | - | 5 | 578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rqcd1| Gene Symbol: Rqcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009357.1 | NP_001009357.1 | 5 | 863 | ||
| GenBank mRNA | BC087134.1 | - | 5 | 863 | 7122272 | |
| FQ225078.1 | - | 5 | 578 | |||