siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173986.1 | NP_001167457.1 | 8, 9 | 1214 | ||
| XM_006245174.2 | XP_006245236.1 | 8, 9 | 1227 | |||
| XM_008767221.1 | XP_008765443.1 | 4, 5 | 636 | |||
| GenBank mRNA | FQ218305.1 | - | 8, 9 | 1216 | ||

Stk36
Genomic Map