siRNA Details
| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173986.1 | NP_001167457.1 | 5, 6 | 699 | ||
| XM_006245174.2 | XP_006245236.1 | 5, 6 | 712 | |||
| GenBank mRNA | FQ218305.1 | - | 5, 6 | 701 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk36| Gene Symbol: Stk36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173986.1 | NP_001167457.1 | 5, 6 | 699 | ||
| XM_006245174.2 | XP_006245236.1 | 5, 6 | 712 | |||
| GenBank mRNA | FQ218305.1 | - | 5, 6 | 701 | ||