siRNA Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 17 | 1714 | ||
| GenBank mRNA | BC079364.1 | - | 17 | 1714 | 7109296 | |
| FQ233375.1 | - | 10 | 1003 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farsb| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 17 | 1714 | ||
| GenBank mRNA | BC079364.1 | - | 17 | 1714 | 7109296 | |
| FQ233375.1 | - | 10 | 1003 | |||