siRNA Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004252.1 | NP_001004252.1 | 12 | 1120 | ||
| XM_006245189.2 | XP_006245251.1 | 12 | 1156 | |||
| GenBank mRNA | BC079364.1 | - | 12 | 1120 | 7109296 | |
| FQ233375.1 | - | 5 | 409 | |||

Farsb
Genomic Map