siRNA Details
| Gene Symbol: Cul3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106923.1 | NP_001100393.1 | 7 | 986 | ||
| XM_008767226.1 | XP_008765448.1 | 8 | 1138 | |||
| XM_008767227.1 | XP_008765449.1 | 7 | 1862 | |||
| XM_008767228.1 | XP_008765450.1 | 7 | 2353 | |||
| XM_008767229.1 | XP_008765451.1 | 7 | 2150 | |||
| GenBank mRNA | BC168969.1 | - | 7 | 991 | 9026830 | |

Cul3
Genomic Map