siRNA Details
| Gene Symbol: Dock10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767310.1 | XP_008765532.1 | 9333 | |||
| XM_008767311.1 | XP_008765533.1 | 9333 | ||||
| XM_008767312.1 | XP_008765534.1 | 9306 | ||||
| XM_008767313.1 | XP_008765535.1 | 9333 | ||||
| XM_008767314.1 | XP_008765536.1 | 9333 | ||||
| XM_008767315.1 | XP_008765537.1 | 9333 | ||||

Dock10
Genomic Map