siRNA Details
Gene Symbol: Olr1344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000483.1 | NP_001000483.1 | 1 | 202 |
Gene Symbol: Olr1343 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001088.1 | NP_001001088.1 | 1 | 202 |
