siRNA Details
| Gene Symbol: Srbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239708.2 | XP_006239770.1 | 14 | 1978 | ||
| XM_237474.7 | XP_237474.5 | 14 | 1847 | |||
| GenBank mRNA | FQ212138.1 | - | 11 | 1448 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srbd1| Gene Symbol: Srbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239708.2 | XP_006239770.1 | 14 | 1978 | ||
| XM_237474.7 | XP_237474.5 | 14 | 1847 | |||
| GenBank mRNA | FQ212138.1 | - | 11 | 1448 | ||