siRNA Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 13 | 1312 | ||
| GenBank mRNA | FN804600.1 | - | 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 13 | 1312 | ||
| GenBank mRNA | FN804600.1 | - | 2 | 241 | ||