siRNA Details
| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 6, 7 | 496 | ||
| GenBank mRNA | FM104860.1 | - | 6, 7 | 432 | ||
| FN805880.1 | - | 4, 5 | 276 | |||
| FN805881.1 | - | 6, 7 | 416 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2| Gene Symbol: Copg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106929.2 | NP_001100399.1 | 6, 7 | 496 | ||
| GenBank mRNA | FM104860.1 | - | 6, 7 | 432 | ||
| FN805880.1 | - | 4, 5 | 276 | |||
| FN805881.1 | - | 6, 7 | 416 | |||