siRNA Details
| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106930.1 | NP_001100400.1 | 3 | 425 | ||
| XM_008768871.1 | XP_008767093.1 | 3 | 456 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Efcab1| Gene Symbol: Efcab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106930.1 | NP_001100400.1 | 3 | 425 | ||
| XM_008768871.1 | XP_008767093.1 | 3 | 456 | |||