siRNA Details
| Gene Symbol: RGD1304704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047874.1 | NP_001041339.1 | 2, 3 | 170 | ||
| GenBank mRNA | AY539943.1 | - | 2, 3 | 170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1304704| Gene Symbol: RGD1304704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047874.1 | NP_001041339.1 | 2, 3 | 170 | ||
| GenBank mRNA | AY539943.1 | - | 2, 3 | 170 | ||