siRNA Details
| Gene Symbol: IFRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006764.4 | NP_006755.4 | 8 | 1163 | ||
| GenBank mRNA | AK097765.1 | - | 1 | 595 | ||
| AK309114.1 | - | 6 | 955 | |||
| BC001327.1 | - | 8 | 990 | 3456988 | ||
| BC001676.2 | - | 8 | 975 | 2963821 | ||
| BC007265.1 | - | 8 | 998 | 3138466 | ||
| BC007437.2 | - | 8 | 1023 | 2961581 | ||
| BT019999.1 | - | 8 | 842 | |||
| U09585.1 | - | 8 | 840 | |||
| Y12395.1 | - | 11 | 1340 | |||

IFRD2
Genomic Map