siRNA Details
| Gene Symbol: Satl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163543.1 | NP_001157015.1 | 4, 5 | 2057 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Satl1| Gene Symbol: Satl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163543.1 | NP_001157015.1 | 4, 5 | 2057 | ||