siRNA Details
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257198.2 | XP_006257260.1 | 9, 10 | 1726 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dach2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257198.2 | XP_006257260.1 | 9, 10 | 1726 |