siRNA Details
| Gene Symbol: RGD1561552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126271.1 | NP_001119743.1 | 2 | 945 | ||
| GenBank mRNA | BC162049.1 | - | 2 | 945 | 7111186 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1561552| Gene Symbol: RGD1561552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126271.1 | NP_001119743.1 | 2 | 945 | ||
| GenBank mRNA | BC162049.1 | - | 2 | 945 | 7111186 | |