siRNA Details
| Gene Symbol: Fam46d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106936.1 | NP_001100406.1 | 1 | 868 | ||
| XM_006257181.2 | XP_006257243.1 | 4 | 1283 | |||
| XM_008773335.1 | XP_008771557.1 | 5 | 1395 | |||
| XM_008773336.1 | XP_008771558.1 | 5 | 1161 | |||
| XM_008773337.1 | XP_008771559.1 | 5 | 1609 | |||
| XM_008773338.1 | XP_008771560.1 | 4 | 1281 | |||

Fam46d
Genomic Map