siRNA Details
| Gene Symbol: Magee2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106941.1 | NP_001100411.1 | 1 | 673 | ||
| XM_006257144.2 | XP_006257206.1 | 3 | 2026 | |||
| XM_008773341.1 | XP_008771563.1 | 2 | 2213 | |||
| XM_008773342.1 | XP_008771564.1 | 3 | 2086 | |||

Magee2
Genomic Map