siRNA Details
| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212518.1 | NP_997683.1 | 1, 2 | 186 | ||
| GenBank mRNA | AJ621255.1 | - | 1, 2 | 186 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb7| Gene Symbol: Abcb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212518.1 | NP_997683.1 | 1, 2 | 186 | ||
| GenBank mRNA | AJ621255.1 | - | 1, 2 | 186 | ||