siRNA Details
| Gene Symbol: Pabpc1l2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257150.2 | XP_006257212.1 | 1 | 559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc1l2a| Gene Symbol: Pabpc1l2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006257150.2 | XP_006257212.1 | 1 | 559 | ||