siRNA Details
| Gene Symbol: Nyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100967.1 | NP_001094437.1 | 3 | 1183 | ||
| GenBank mRNA | DQ393414.1 | - | 3 | 1183 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nyx| Gene Symbol: Nyx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100967.1 | NP_001094437.1 | 3 | 1183 | ||
| GenBank mRNA | DQ393414.1 | - | 3 | 1183 | ||