siRNA Details
| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106955.2 | NP_001100425.1 | 2, 3 | 280 | ||
| GenBank mRNA | BC161862.1 | - | 2, 3 | 280 | 7097051 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbc1d25| Gene Symbol: Tbc1d25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106955.2 | NP_001100425.1 | 2, 3 | 280 | ||
| GenBank mRNA | BC161862.1 | - | 2, 3 | 280 | 7097051 | |