siRNA Details
| Gene Symbol: Wdr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013958.1 | NP_001013980.1 | 10 | 861 | ||
| XM_006256710.2 | XP_006256772.1 | 10 | 1033 | |||
| XM_006256712.2 | XP_006256774.1 | 10 | 1144 | |||
| XM_006256713.2 | XP_006256775.1 | 10 | 918 | |||
| XM_006256714.2 | XP_006256776.1 | 10 | 1038 | |||
| XM_008773071.1 | XP_008771293.1 | 10 | 931 | |||
| GenBank mRNA | BC085816.1 | - | 10 | 861 | 7123612 | |
| FQ225212.1 | - | 5 | 525 | |||
| FQ232559.1 | - | 5 | 525 | |||
| FQ233394.1 | - | 5 | 525 | |||

Wdr45
Genomic Map