siRNA Details
| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207601.1 | NP_997484.1 | 2 | 248 | ||
| GenBank mRNA | BC061844.1 | - | 2 | 248 | 5621135 | |
| FQ220564.1 | - | 2 | 287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plp2| Gene Symbol: Plp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207601.1 | NP_997484.1 | 2 | 248 | ||
| GenBank mRNA | BC061844.1 | - | 2 | 248 | 5621135 | |
| FQ220564.1 | - | 2 | 287 | |||