siRNA Details
| Gene Symbol: Nxf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191731.2 | NP_001178660.2 | 8 | 966 | ||
| XM_006257271.2 | XP_006257333.1 | 10 | 1093 | |||
| XM_008773409.1 | XP_008771631.1 | 9 | 1023 | |||
| XM_008773410.1 | XP_008771632.1 | 10 | 1149 | |||
| XM_008773411.1 | XP_008771633.1 | 10 | 1146 | |||
| XM_008773412.1 | XP_008771634.1 | 10 | 1026 | |||

Nxf3
Genomic Map