siRNA Details
| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191618.2 | NP_001178547.1 | 5 | 1286 | ||
| XM_006257038.2 | XP_006257100.1 | 5 | 1437 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspyl2| Gene Symbol: Tspyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191618.2 | NP_001178547.1 | 5 | 1286 | ||
| XM_006257038.2 | XP_006257100.1 | 5 | 1437 | |||