siRNA Details
| Gene Symbol: LOC102557137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773237.1 | XP_008771459.1 | 1 | 985 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102557137| Gene Symbol: LOC102557137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773237.1 | XP_008771459.1 | 1 | 985 | ||