siRNA Details
| Gene Symbol: Frmpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106960.1 | NP_001100430.1 | 8 | 816 | ||
| XM_008773161.1 | XP_008771383.1 | 8 | 898 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmpd4| Gene Symbol: Frmpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106960.1 | NP_001100430.1 | 8 | 816 | ||
| XM_008773161.1 | XP_008771383.1 | 8 | 898 | |||