siRNA Details
Gene Symbol: Asb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106962.1 | NP_001100432.1 | 5 | 505 | ||
XM_006256858.2 | XP_006256920.1 | 5 | 859 | |||
XM_008773166.1 | XP_008771388.1 | 5 | 910 | |||
GenBank mRNA | FQ216453.1 | - | 5 | 611 | ||
FQ224911.1 | - | 5 | 564 |
