siRNA Details
| Gene Symbol: Mbtps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035007.1 | NP_001030179.1 | 5, 6 | 756 | ||
| XM_006256944.2 | XP_006257006.1 | 3, 4 | 445 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_172307, AK148325, BC038343, AK164486.

Mbtps2
Genomic Map