siRNA Details
| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106964.1 | NP_001100434.1 | 347 | |||
| GenBank mRNA | BC167084.1 | - | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbl1x| Gene Symbol: Tbl1x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106964.1 | NP_001100434.1 | 347 | |||
| GenBank mRNA | BC167084.1 | - | 442 | |||