siRNA Details
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106964.1 | NP_001100434.1 | 712 | |||
GenBank mRNA | BC167084.1 | - | 807 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbl1x | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106964.1 | NP_001100434.1 | 712 | |||
GenBank mRNA | BC167084.1 | - | 807 |