siRNA Details
| Gene Symbol: Mageb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256990.2 | XP_006257052.1 | 1 | 310 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mageb2| Gene Symbol: Mageb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006256990.2 | XP_006257052.1 | 1 | 310 | ||