siRNA Details
| Gene Symbol: Rbm31y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_229073.3 | XP_229073.1 | 1 | 1528 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbm31y| Gene Symbol: Rbm31y | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_229073.3 | XP_229073.1 | 1 | 1528 | ||