siRNA Details
| Gene Symbol: Actrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013961.1 | NP_001013983.1 | 1 | 757 | ||
| GenBank mRNA | BC083679.1 | - | 1 | 757 | 7135866 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actrt1| Gene Symbol: Actrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013961.1 | NP_001013983.1 | 1 | 757 | ||
| GenBank mRNA | BC083679.1 | - | 1 | 757 | 7135866 | |