siRNA Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257541.2 | XP_006257603.1 | 1776 | |||
XM_006257542.2 | XP_006257604.1 | 1675 | ||||
XM_008773539.1 | XP_008771761.1 | 15 | 1663 | |||
XM_008773540.1 | XP_008771762.1 | 1632 | ||||
XM_008773541.1 | XP_008771763.1 | 14 | 1627 | |||
XM_008773542.1 | XP_008771764.1 | 13 | 2657 |
