siRNA Details
| Gene Symbol: DEK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134709.1 | NP_001128181.1 | 5 | 602 | ||
| NM_003472.3 | NP_003463.1 | 6 | 704 | |||
| XM_017011273.1 | XP_016866762.1 | 6 | 676 | |||
| GenBank mRNA | AK294079.1 | - | 6 | 587 | ||
| AK297749.1 | - | 5 | 575 | |||
| AK298089.1 | - | 6 | 616 | |||
| AK312616.1 | - | 6 | 633 | |||
| BC035259.1 | - | 6 | 676 | 5122743 | ||
| X64229.1 | - | 6 | 544 | |||
 Back To Top
 Back To Top
 DEK
DEK Genomic Map
Genomic Map Back To Top
 Back To Top