siRNA Details
| Gene Symbol: Igsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175763.2 | NP_786939.1 | 18 | 3757 | ||
| XM_006257557.2 | XP_006257619.1 | 19 | 3878 | |||
| XM_006257558.1 | XP_006257620.1 | 18 | 3828 | |||
| XM_006257559.1 | XP_006257621.1 | 18 | 3744 | |||
| XM_006257560.1 | XP_006257622.1 | 18 | 3831 | |||
| XM_006257561.1 | XP_006257623.1 | 17 | 3543 | |||
| XM_008773563.1 | XP_008771785.1 | 18 | 3831 | |||
| XM_008773564.1 | XP_008771786.1 | 11 | 2834 | |||
| GenBank mRNA | AF322216.1 | - | 18 | 3757 | ||

Igsf1
Genomic Map