siRNA Details
| Gene Symbol: Magea11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013962.1 | NP_001013984.1 | 3 | 961 | ||
| GenBank mRNA | BC083676.1 | - | 3 | 961 | 7135789 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Magea11| Gene Symbol: Magea11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013962.1 | NP_001013984.1 | 3 | 961 | ||
| GenBank mRNA | BC083676.1 | - | 3 | 961 | 7135789 | |