siRNA Details
| Gene Symbol: Mmgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106970.1 | NP_001100440.1 | 4 | 448 | ||
| GenBank mRNA | BC168927.1 | - | 4 | 369 | 7108747 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmgt1| Gene Symbol: Mmgt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106970.1 | NP_001100440.1 | 4 | 448 | ||
| GenBank mRNA | BC168927.1 | - | 4 | 369 | 7108747 | |