siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013963.1 | NP_001013985.1 | 4 | 928 | ||
| GenBank mRNA | BC081991.1 | - | 4 | 928 | 7129542 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpped1| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013963.1 | NP_001013985.1 | 4 | 928 | ||
| GenBank mRNA | BC081991.1 | - | 4 | 928 | 7129542 | |