siRNA Details
| Gene Symbol: Rbfox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106974.2 | NP_001100444.2 | 5, 6 | 852 | ||
| XM_006245774.2 | XP_006245836.1 | 5, 6 | 1386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbfox1| Gene Symbol: Rbfox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106974.2 | NP_001100444.2 | 5, 6 | 852 | ||
| XM_006245774.2 | XP_006245836.1 | 5, 6 | 1386 | |||