siRNA Details
| Gene Symbol: Anks3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009676.1 | NP_001009676.1 | 3 | 289 | ||
| XM_006245788.1 | XP_006245850.1 | 3 | 332 | |||
| XM_006245789.2 | XP_006245851.1 | 3 | 332 | |||
| XM_006245790.1 | XP_006245852.1 | 3 | 225 | |||
| XM_008767482.1 | XP_008765704.1 | 3 | 332 | |||
| XM_008767483.1 | XP_008765705.1 | 2 | 2061 | |||
| GenBank mRNA | BC087062.1 | - | 3 | 289 | 7131099 | |

Anks3
Genomic Map