siRNA Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013964.1 | NP_001013986.1 | 1, 2 | 221 | ||
| XM_006245792.2 | XP_006245854.1 | 1, 2 | 225 | |||
| XM_006245794.2 | XP_006245856.1 | 1, 2 | 224 | |||
| XM_006245796.2 | XP_006245858.1 | 1, 2 | 225 | |||
| GenBank mRNA | BC083621.1 | - | 1, 2 | 221 | 7134018 | |

Mgrn1
Genomic Map