siRNA Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013964.1 | NP_001013986.1 | 2 | 308 | ||
| XM_006245792.2 | XP_006245854.1 | 2 | 312 | |||
| XM_006245793.2 | XP_006245855.1 | 2 | 309 | |||
| XM_006245794.2 | XP_006245856.1 | 2 | 311 | |||
| XM_006245795.2 | XP_006245857.1 | 2 | 308 | |||
| XM_006245796.2 | XP_006245858.1 | 2 | 312 | |||
| XM_008767485.1 | XP_008765707.1 | 2 | 308 | |||
| XM_008767486.1 | XP_008765708.1 | 2 | 308 | |||
| GenBank mRNA | BC083621.1 | - | 2 | 308 | 7134018 | |
| HE574489.1 | - | 2 | 172 | |||
| HE574490.1 | - | 2 | 172 | |||

Mgrn1
Genomic Map